LIF372158365 Life Chemicals Inc.

{4-[(4-Fluorobenzyl)oxy]phenyl}amine

  • MDL number MFCD08056129
  • Cas Number : 53234-85-8
  • Molecular Weight : 217.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(4-fluorobenzyl)oxy]aniline
Inchi Code 1S/C13H12FNO/c14-11-3-1-10(2-4-11)9-16-13-7-5-12(15)6-8-13/h1-8H,9,15H2
Inchi Key WKJLZHPOOMPGPI-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart