LIF372158391 Life Chemicals Inc.

methyl 3-(3-chlorophenyl)-1H-pyrazole-5-carboxylate

  • MDL number MFCD12188758
  • Cas Number : 1187360-62-8
  • Molecular Weight : 236.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(3-chlorophenyl)-1H-pyrazole-5-carboxylate
Inchi Code 1S/C11H9ClN2O2/c1-16-11(15)10-6-9(13-14-10)7-3-2-4-8(12)5-7/h2-6H,1H3,(H,13,14)
Inchi Key CKYXRVBZEALSPS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart