LIF372158398 Life Chemicals Inc.

2-chloro-1-methyl-1H-benzimidazole

  • MDL number MFCD00769821
  • Cas Number : 1849-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart