LIF372158400 Life Chemicals Inc.

5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00608153
  • Cas Number : 40704-11-8
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart