LIF372158409 Life Chemicals Inc.

3-(4-fluorophenoxy)benzoic acid

  • MDL number MFCD03840105
  • Cas Number : 129623-62-7
  • Molecular Weight : 232.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-fluorophenoxy)benzoic acid
Inchi Code 1S/C13H9FO3/c14-10-4-6-11(7-5-10)17-12-3-1-2-9(8-12)13(15)16/h1-8H,(H,15,16)
Inchi Key JEZVFQIMFSGLFL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart