LIF372158418 Life Chemicals Inc.

1,2,3,5,6,7,8,9-octahydro-4H-cyclopenta[c]quinolin-4-one

  • MDL number MFCD22613713
  • Cas Number : 59663-81-9
  • Molecular Weight : 189.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3,6,7,8,9-hexahydro-1H-cyclopenta[c]quinolin-4-ol
Inchi Code 1S/C12H15NO/c14-12-10-6-3-5-8(10)9-4-1-2-7-11(9)13-12/h1-7H2,(H,13,14)
Inchi Key YPDWWSMFSQNAFV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart