LIF372158419 Life Chemicals Inc.

6-methyl-1,4-dihydropyrido[2,3-b]pyrazine-2,3-dione

  • MDL number MFCD22613714
  • Cas Number : 144435-06-3
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methylpyrido[2,3-b]pyrazine-2,3-diol
Inchi Code 1S/C8H7N3O2/c1-4-2-3-5-6(9-4)11-8(13)7(12)10-5/h2-3H,1H3,(H,10,12)(H,9,11,13)
Inchi Key WWJQZVUJFGDORP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart