LIF372158422 Life Chemicals Inc.

1H,2H,3H,4H-pyrido[2,3-b]pyrazine

  • MDL number MFCD08062758
  • Cas Number : 35808-40-3
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart