LIF372158426 Life Chemicals Inc.

1H,2H,3H,5H,6H,7H,8H-cyclopenta[b]quinolin-9-amine

  • MDL number MFCD00618424
  • Cas Number : 62732-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart