LIF372158427 Life Chemicals Inc.

pyrido[2,3-b]pyrazine

  • MDL number MFCD00006717
  • Cas Number : 322-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart