LIF372158428 Life Chemicals Inc.

N-methyl-3-nitropyridin-2-amine

  • MDL number MFCD00223335
  • Cas Number : 4093-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart