LIF372158436 Life Chemicals Inc.

5-(4-tert-butylphenyl)-1,2,4-thiadiazol-3-amine

  • MDL number MFCD11655562
  • Cas Number : 1086385-86-5
  • Molecular Weight : 233.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(4-tert-butylphenyl)-1,2,4-thiadiazol-3-ylamine
Inchi Code 1S/C12H15N3S/c1-12(2,3)9-6-4-8(5-7-9)10-14-11(13)15-16-10/h4-7H,1-3H3,(H2,13,15)
Inchi Key KMTRRQKJAQTEEY-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart