LIF372158441 Life Chemicals Inc.

1-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethan-1-one

  • MDL number MFCD00179440
  • Cas Number : 84162-82-3
  • Molecular Weight : 267.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart