LIF372158444 Life Chemicals Inc.

3-fluoro-5-nitrobenzoic acid

  • MDL number MFCD05864360
  • Cas Number : 14027-75-9
  • Molecular Weight : 185.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart