LIF372158448 Life Chemicals Inc.

ethyl 3-amino-4-nitrobenzoate

  • MDL number MFCD14584886
  • Cas Number : 84228-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart