LIF372158449 Life Chemicals Inc.

ethyl 1H-benzimidazole-5-carboxylate

  • MDL number MFCD09027455
  • Cas Number : 58842-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart