LIF372158450 Life Chemicals Inc.

3-amino-4-nitrobenzamide

  • MDL number MFCD14584887
  • Cas Number : 131508-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart