LIF372158454 Life Chemicals Inc.

4-[(diethoxyphosphoryl)methyl]benzoic acid

  • MDL number MFCD00440869
  • Cas Number : 28149-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart