LIF372158456 Life Chemicals Inc.

6-amino-3-(2-fluorobenzyl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD11655575
  • Cas Number : 639780-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart