LIF372158461 Life Chemicals Inc.

diethyl {5-[2-(3-methylbutyl)-1,3-dithian-2-yl]-2-furyl}phosphonate

  • MDL number MFCD14584896
  • Cas Number : 1199589-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart