LIF372158464 Life Chemicals Inc.

[5-(2-amino-5-isobutyl-1,3-thiazol-4-yl)-2-furyl]phosphonic acid

  • MDL number MFCD14584901
  • Cas Number : 261365-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart