LIF372158466 Life Chemicals Inc.

5-nitro-1,3-benzothiazole

  • MDL number MFCD00085891
  • Molecular Weight : 180.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart