LIF372158468 Life Chemicals Inc.

(6-bromo-2-methoxyquinolin-3-yl)(3-chloro-4-fluorophenyl)methanol

  • MDL number MFCD14584908
  • Cas Number : 1199589-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart