LIF372158469 Life Chemicals Inc.

tert-butyl [2-cyclobutyl-1-(hydroxymethyl)ethyl]carbamate

  • MDL number MFCD14584910
  • Cas Number : 816429-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart