LIF372158472 Life Chemicals Inc.

N-[(tert-butylamino)carbonyl]-3-methylvaline

  • MDL number MFCD12176580
  • Cas Number : 1155278-31-1
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart