LIF372158479 Life Chemicals Inc.

N~1~-[3-(dimethylamino)propyl]-N~1~-methyl-4-(trifluoromethyl)benzene-1,2-diamine

  • MDL number MFCD14584932
  • Cas Number : 861881-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart