LIF372158489 Life Chemicals Inc.

4-chloro-2-(4-fluorophenyl)quinazoline

  • MDL number MFCD04115224
  • Cas Number : 113242-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart