LIF372158490 Life Chemicals Inc.

N-(3-methoxyphenyl)-3-oxobutanamide

  • MDL number MFCD01001275
  • Cas Number : 25233-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart