LIF372158491 Life Chemicals Inc.

2-chloroisophthalic acid

  • MDL number MFCD00156972
  • Cas Number : 13049-16-6
  • Molecular Weight : 200.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart