LIF372158493 Life Chemicals Inc.

2-(3-aminoimidazo[1,2-a]pyridin-2-yl)-4-bromophenol

  • MDL number MFCD02658347
  • Cas Number : 80493-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart