LIF372158496 Life Chemicals Inc.

1-(chloroacetyl)-4-(2-fluorophenyl)piperazine

  • MDL number MFCD01480897
  • Cas Number : 546116-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart