LIF372158497 Life Chemicals Inc.

4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride

  • MDL number MFCD00024882
  • Cas Number : 32333-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart