LIF372158498 Life Chemicals Inc.

4-amino-1-(3,4-dihydroxy-5-(hydroxymethyl)-3-methyltetrahydrofuran-2-yl)pyrimidin-2(1H)-one

  • MDL number MFCD16653460

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart