LIF372158502 Life Chemicals Inc.

3-(4-fluorophenoxy)benzaldehyde

  • MDL number MFCD02258991
  • Cas Number : 65295-61-6
  • Molecular Weight : 216.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart