LIF372158510 Life Chemicals Inc.

2-chloro-N-(3-methylphenyl)acetamide

  • MDL number MFCD00018901
  • Cas Number : 32428-61-8
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart