LIF372158511 Life Chemicals Inc.

3-chloro-N-(4-ethoxyphenyl)propanamide

  • MDL number MFCD00087032
  • Cas Number : 19314-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart