LIF372158514 Life Chemicals Inc.

N-(4-tert-butylphenyl)-3-chloropropanamide

  • MDL number MFCD06409310
  • Cas Number : 20330-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart