LIF372158517 Life Chemicals Inc.

N-(1,3-benzodioxol-5-ylmethyl)-3-chloropropanamide

  • MDL number MFCD03362889
  • Cas Number : 95639-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart