LIF372158519 Life Chemicals Inc.

4-ethyl-2-mercapto-7,7-dimethyl-5-oxo-5,6,7,8-tetrahydroquinoline-3-carbonitrile

  • MDL number MFCD00435842
  • Cas Number : 885268-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart