LIF372158524 Life Chemicals Inc.

methyl 2-amino-4-(3,4-dimethylphenyl)thiophene-3-carboxylate

  • MDL number MFCD01922223
  • Cas Number : 350990-08-8
  • Molecular Weight : 261.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart