LIF372158531 Life Chemicals Inc.

methyl 2-amino-4-(4-methoxyphenyl)thiophene-3-carboxylate

  • MDL number MFCD01859807
  • Cas Number : 350988-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart