LIF372158534 Life Chemicals Inc.

2-chloro-1-[1-(2,5-dichlorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]ethanone

  • MDL number MFCD06409342
  • Cas Number : 885268-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart