LIF372158541 Life Chemicals Inc.

hydroxy(3-hydroxy-4-methoxyphenyl)acetic acid

  • MDL number MFCD00004230
  • Cas Number : 3695-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart