LIF372158543 Life Chemicals Inc.

4'-chlorobiphenyl-4-carbaldehyde

  • MDL number MFCD01631911
  • Cas Number : 80565-30-6
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart