LIF372158555 Life Chemicals Inc.

1,3-benzothiazole-6-carbonitrile

  • MDL number MFCD06659656
  • Cas Number : 58249-61-9
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart