LIF372158560 Life Chemicals Inc.

2-methyl-6-propylphenol

  • MDL number MFCD00086137
  • Cas Number : 3520-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart