LIF372158561 Life Chemicals Inc.

2-morpholin-4-ylnicotinaldehyde

  • MDL number MFCD03086195
  • Cas Number : 465514-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart