LIF372158563 Life Chemicals Inc.

1-ethynyl-4-(propan-2-yl)benzene

  • MDL number MFCD03839988
  • Cas Number : 23152-99-0
  • Molecular Weight : 144.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart