LIF372158565 Life Chemicals Inc.

1H-pyrazolo[3,4-d]pyrimidine-4-thiol

  • MDL number MFCD00005687
  • Cas Number : 5334-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart