LIF372158570 Life Chemicals Inc.

(4-bromobutyl)cyclohexane

  • MDL number MFCD06658371
  • Cas Number : 60439-16-9
  • Molecular Weight : 219.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart